Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Isopropylidenemalononitrile ≥98.0%

Supplier: Decon Laboratories Ltd

I0488-25G I0488-500G
TCI0488-025GEA 103 USD InStock
TCI0488-025G TCI0488-500G
Isopropylidenemalononitrile ≥98.0%
Isopropylidenemalononitrile
CAS Number: 13166-10-4
MDL Number: MFCD00082572
Molecular Formula: C6H6N2
Molecular Weight: 106.13
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 112
Flash Point (°C): 101
Specific Gravity (20/20): 0.96
Storage Temperature: 0-10°C

Other Product Names / Synonyms: 1,1-Dicyano-2-methyl-1-propene


Formula: C₆H₆N₂
Boiling Pt: 112 °C
Density: 0.96 (20 °C)
Flash Pt: 101 °C
MDL Number: MFCD00082572
CAS Number: 13166-10-4
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1eb1cebbaca09d49c401aef220f07793076da1ec&vid=1eb1cebbaca09d49c401aef220f07793076da1ec&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty