Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-(-)-4-Isopropyl-3-propionyl-2-oxazolidinone ≥98.0%

Supplier: Decon Laboratories Ltd

I0594-5G I0594-1G
TCI0594-5GEA 161.1 USD InStock
TCI0594-5G TCI0594-1G
(R)-(-)-4-Isopropyl-3-propionyl-2-oxazolidinone ≥98.0%
(R)-(-)-4-Isopropyl-3-propionyl-2-oxazolidinone
CAS Number: 89028-40-0
MDL Number: MFCD00269691
Molecular Formula: C9H15NO3
Molecular Weight: 185.22
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.10
Specific rotation [a]20/D: -90 deg (neat)

Product Categories: Synthetic Organic Chemistry, Chiral Auxiliaries
Other Product Names / Synonyms: (R)-(-)-3-Propionyl-4-isopropyl-2-oxazolidinone


Formula: C₉H₁₅NO₃
Density: 1.10 (20 °C)
MDL Number: MFCD00269691
CAS Number: 89028-40-0
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=19455b9c10d3f045c89a1af0a4ced96c9c62bd4c&vid=19455b9c10d3f045c89a1af0a4ced96c9c62bd4c&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty