Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Isopropyl 2-methylbutyrate ≥98.0%

Supplier: Decon Laboratories Ltd

I0736-25ML I0736-500ML
TCI0736-25MLEA 34 USD InStock
TCI0736-25ML TCI0736-500ML
Isopropyl 2-methylbutyrate ≥98.0%
Isopropyl 2-methylbutyrate
CAS Number: 66576-71-4
MDL Number: MFCD00085203
Molecular Formula: C8H16O2
Molecular Weight: 144.21
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 144
Flash Point (°C): 32
Specific Gravity (20/20): 0.85

Other Product Names / Synonyms: 2-Methylbutyric Acid Isopropyl Ester


Formula: C₈H₁₆O₂
Boiling Pt: 144 °C
Density: 0.85 (20 °C)
Flash Pt: 32 °C
MDL Number: MFCD00085203
CAS Number: 66576-71-4
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e60cabcf129a5b2f219f5941bfe12913bc35ac27&vid=e60cabcf129a5b2f219f5941bfe12913bc35ac27&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty