Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl (±)-lactate ≥98.0% for synthesis

Supplier: Decon Laboratories Ltd
Danger

L0004-500ML L0004-25ML
TCL0004-500MLEA 180.03 USD InStock
TCL0004-500ML TCL0004-025ML
Methyl (±)-lactate ≥98.0% for synthesis
Methyl (±)-lactate
CAS Number: 547-64-8
MDL Number: MFCD00066367
Molecular Formula: C4H8O3
Molecular Weight: 104.11
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 144
Flash Point (°C): 54
Specific Gravity (20/20): 1.09

Other Product Names / Synonyms: Lactic Acid Methyl Ester


Formula: C₄H₈O₃
MW: 104.11 g/mol
Boiling Pt: 35
Density: 1.09 (20 °C)
Flash Pt: 54 °C
MDL Number: MFCD00066367
CAS Number: 547-64-8
EINECS: 208-930-0
UN: 3272
ADR: 3 III
Merck Index: 12,06167
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=73d0cfa7773853ae506eef3631b80cec84dd5c74&vid=73d0cfa7773853ae506eef3631b80cec84dd5c74&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty