Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Lacmoid

Supplier: Decon Laboratories Ltd

L0079-1G L0079-5G
TCL0079-001GEA 33.56 USD InStock
TCL0079-001G TCL0079-005G
Lacmoid
Lacmoid
CAS Number: 42249-61-6
MDL Number: MFCD00152579
Form: Crystal
Color: Black

Other Product Names / Synonyms: Resorcinol Blue


MDL Number: MFCD00152579
CAS Number: 42249-61-6
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty