Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Loperamide hydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: Decon Laboratories Ltd

L0154-25G L0154-5G
TCL0154-025GEA 297 USD InStock
TCL0154-025G TCL0154-005G
Loperamide hydrochloride ≥98.0% (by HPLC, titration analysis)
Loperamide hydrochloride
CAS Number: 34552-83-5 MDL Number: MFCD00058581 Molecular Formula: C29H33ClN2O2 Molecular Weight: 513.50 Purity/Analysis Method: 98.0% (T,HPLC) Form: Crystal Melting point (°C): 223


Formula: C₂₉H₃₃ClN₂O₂
Melting Pt: 223 °C
MDL Number: MFCD00058581
CAS Number: 34552-83-5
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=669029c6366ce97f4ceef7169ad06b2ce9138f53&vid=669029c6366ce97f4ceef7169ad06b2ce9138f53&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (T,HPLC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty