Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Lovastatin ≥98.0%

Supplier: Decon Laboratories Ltd

L0214-5G L0214-25G
TCL0214-5GEA 92.71 USD InStock
TCL0214-5G TCL0214-25G
Lovastatin ≥98.0%
Lovastatin
CAS Number: 75330-75-5
MDL Number: MFCD00072164
Molecular Formula: C24H36O5
Molecular Weight: 404.55
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 179
Specific rotation [a]20/D: 320 deg (C=0.5, CH3CN)

Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Inhibitors (for Pharmacological Research), Antilipemic Ingredients, Prodrug Ingredients
Other Product Names / Synonyms: Mevinolin, 6alpha-Methylcompactin, Monacolin K


Formula: C₂₄H₃₆O₅
MW: 404.55 g/mol
Melting Pt: 179 °C
MDL Number: MFCD00072164
CAS Number: 75330-75-5
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=101188846c09237ab0267a1a5b9d82fce51f8fcd&vid=101188846c09237ab0267a1a5b9d82fce51f8fcd&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (HPLC)

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty