Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dodecyl methacrylate ≥97.0% stabilised for synthesis

Supplier: Decon Laboratories Ltd

M0083-500ML M0083-25ML
TCM0083-500MLEA 74.18 USD InStock
TCM0083-500ML TCM0083-025ML
Dodecyl methacrylate ≥97.0% stabilised for synthesis
Dodecyl methacrylate
(stabilized with MEHQ)
CAS Number: 142-90-5
MDL Number: MFCD00008972
Molecular Formula: C16H30O2
Molecular Weight: 254.41
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 167
Flash Point (°C): 143
Specific Gravity (20/20): 0.87

Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Lauryl Methacrylate, Methacrylic Acid Dodecyl Ester, Methacrylic Acid Lauryl Ester


Formula: C₁₆H₃₀O₂
MW: 254.41 g/mol
Boiling Pt: 167 °C
Density: 0.87 (20 °C)
Flash Pt: 143 °C
MDL Number: MFCD00008972
CAS Number: 142-90-5
EINECS: 205-570-6
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=24f2dab29e9b6568e0f709879ede4b8c5ae9ffff&vid=24f2dab29e9b6568e0f709879ede4b8c5ae9ffff&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >97.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty