Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl mercaptan sodium salt ∼15% in water

Supplier: Decon Laboratories Ltd

M0096-500G M0096-25G
TCM0096-500GEA 150.6 USD InStock
TCM0096-500G TCM0096-025G
Methyl mercaptan sodium salt ∼15% in water
Methyl mercaptan sodium salt
CAS Number: 5188-07-8
MDL Number: MFCD00174316
Molecular Formula: CH3NaS
Molecular Weight: 70.08
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 50

Other Product Names / Synonyms: Methanethiol Sodium Salt, Sodium Methanethiolate, Sodium Thiomethoxide


Formula: CH₃NaS
MW: 70.08 g/mol
Flash Pt: 50 °C
MDL Number: MFCD00174316
CAS Number: 5188-07-8
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b6826e2696173fac0a51d61e304e07e9bde88b3a&vid=b6826e2696173fac0a51d61e304e07e9bde88b3a&prid=web&clid=SAPDAM
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty