Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(±)-3-Methylcyclohexanone ≥97.0%

Supplier: Decon Laboratories Ltd

M0198-500ML M0198-25ML M0198-100ML
TCM0198-500MLEA 218 USD InStock
TCM0198-500ML TCM0198-025ML TCM0198-100ML
(±)-3-Methylcyclohexanone ≥97.0%
(±)-3-Methylcyclohexanone
CAS Number: 591-24-2
MDL Number: MFCD00001639
Molecular Formula: C7H12O
Molecular Weight: 112.17
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 169
Flash Point (°C): 48
Specific Gravity (20/20): 0.92


Formula: C₇H₁₂O
Boiling Pt: 169 °C
Melting Pt: -73 °C
Density: 0.92 (20 °C)
Flash Pt: 48 °C
Storage Temperature: Ambient
MDL Number: MFCD00001639
CAS Number: 591-24-2
EINECS: 209-710-7
UN: 1993
ADR: 3,III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bbfec81fbceb27a4eff56224a89b8b207617c4ad&vid=bbfec81fbceb27a4eff56224a89b8b207617c4ad&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >97.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty