Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Methylcytosine hydrochloride ≥98.0% (by HPLC)

Supplier: Decon Laboratories Ltd

M0204-1G M0204-5G M0204-100MG
TCM0204-001GEA 68 USD InStock
TCM0204-001G TCM0204-5G TCM0204-100MG
5-Methylcytosine hydrochloride ≥98.0% (by HPLC)
5-Methylcytosine hydrochloride
CAS Number: 58366-64-6
MDL Number: MFCD00035313
Molecular Formula: C5H7N3O
Molecular Weight: 161.59
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal


Formula: C₅H₇N₃O
MDL Number: MFCD00035313
CAS Number: 58366-64-6
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=33fd37ca4dfe598dd523d5f2b68413acd0374a56&vid=33fd37ca4dfe598dd523d5f2b68413acd0374a56&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (HPLC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty