Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-3-amino-2-methylbenzoate ≥98.0%

Supplier: Decon Laboratories Ltd

A1563-5G A1563-25G
TCA1563-5GEA 80 USD InStock
TCA1563-5G TCA1563-25G
Methyl-3-amino-2-methylbenzoate ≥98.0%
Methyl 3-amino-2-methylbenzoate
CAS Number: 18583-89-6
MDL Number: MFCD00211328
Molecular Formula: C9H11NO2
Molecular Weight: 165.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 160
Flash Point (°C): 104
Specific Gravity (20/20): 1.15

Other Product Names / Synonyms: 3-Amino-2-methylbenzoic Acid Methyl Ester, Methyl 3-Amino-o-toluate, 3-Amino-o-toluic Acid Methyl Ester


Formula: C₉H₁₁NO₂
Boiling Pt: 160 °C
Density: 1.15 (20 °C)
Flash Pt: 104 °C
MDL Number: MFCD00211328
CAS Number: 18583-89-6
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bbaf427caef4fee82c1debace76b1be8d98cdca8&vid=bbaf427caef4fee82c1debace76b1be8d98cdca8&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty