Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Methylmethacrylamide ≥98.0% (by titrimetric analysis) stabilized

Supplier: Decon Laboratories Ltd

M0800-25ML M0800-5ML
TCM0800-025MLEA 180 USD InStock
TCM0800-025ML TCM0800-5ML
N-Methylmethacrylamide ≥98.0% (by titrimetric analysis) stabilized
N-Methylmethacrylamide
(stabilized with HQ)
CAS Number: 3887-02-3
MDL Number: MFCD00048115
Molecular Formula: C5H9NO
Molecular Weight: 99.13
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Boiling point (°C): 110
Specific Gravity (20/20): 0.97
Storage Temperature: 0-10°C


Formula: C₅H₉NO
Boiling Pt: 110 °C
Density: 0.97 (20 °C)
MDL Number: MFCD00048115
CAS Number: 3887-02-3
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=25ff1fbb0e9e78642543c6f097c4163fb1b43f77&vid=25ff1fbb0e9e78642543c6f097c4163fb1b43f77&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (T)

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty