Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methyl-3-butenenitrile ≥75.0%

Supplier: Decon Laboratories Ltd

M0840-500ML M0840-25ML
TCM0840-500MLEA 124 USD InStock
TCM0840-500ML TCM0840-025ML
2-Methyl-3-butenenitrile ≥75.0%
2-Methyl-3-butenenitrile
CAS Number: 16529-56-9
MDL Number: MFCD00042634
Molecular Formula: C5H7N
Molecular Weight: 81.12
Purity/Analysis Method: >75.0% (GC)
Form: Clear Liquid
Boiling point (°C): 124
Flash Point (°C): 15
Specific Gravity (20/20): 0.82

Other Product Names / Synonyms: 3-Cyano-1-butene, 1-Methyl-2-propenyl Cyanide


Formula: C₅H₇N
Boiling Pt: 124 °C
Density: 0.82 (20 °C)
Flash Pt: 15 °C
MDL Number: MFCD00042634
CAS Number: 16529-56-9
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0266c9d93bf07178ab68c19b2d32efccd20094c5&vid=0266c9d93bf07178ab68c19b2d32efccd20094c5&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >75.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty