Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Methoxy-1-trimethylsilyloxypropene ≥95.0%

Supplier: Decon Laboratories Ltd

M1199-5G M1199-1G
TCM1199-5GEA 533.5 USD InStock
TCM1199-5G TCM1199-001G
1-Methoxy-1-trimethylsilyloxypropene ≥95.0%
1-Methoxy-1-trimethylsilyloxypropene
CAS Number: 34880-70-1
MDL Number: MFCD00671554
Molecular Formula: C7H16O2Si
Molecular Weight: 160.29
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 52
Specific Gravity (20/20): 0.87
Storage Temperature: 0-10°C

Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis), Materials Chemistry, Precise Polymerization Reagents
Other Product Names / Synonyms: [(1-Methoxy-1-propenyl)oxy]trimethylsilane


Formula: C₇H₁₆O₂Si
Boiling Pt: 52 °C
Density: 0.87 (20 °C)
MDL Number: MFCD00671554
CAS Number: 34880-70-1
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4307a1377aaacbbe57f543cf9a243c4d8ff58bac&vid=4307a1377aaacbbe57f543cf9a243c4d8ff58bac&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >95.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty