Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methyl-1-naphthoic acid ≥98.0%

Supplier: Decon Laboratories Ltd

M1203-25G M1203-5G
TCM1203-25GEA 95.2 USD InStock
TCM1203-25G TCM1203-005G
4-Methyl-1-naphthoic acid ≥98.0%
4-Methyl-1-naphthoic acid
CAS Number: 4488-40-8
MDL Number: MFCD00671557
Molecular Formula: C12H10O2
Molecular Weight: 186.21
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 178

Other Product Names / Synonyms: 4-Methyl-1-naphthalenecarboxylic Acid


Formula: C₁₂H₁₀O₂
MW: 186.21 g/mol
Melting Pt: 178 °C
MDL Number: MFCD00671557
CAS Number: 4488-40-8
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2b8e3230855bc1cfebadcfd905763a851e48c5e0&vid=2b8e3230855bc1cfebadcfd905763a851e48c5e0&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty