Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(+)-Menthyl chloroformate ≥97.0% (by titrimetric analysis)

Supplier: Decon Laboratories Ltd

M1221-25ML M1221-5ML
TCM1221-025MLEA 213.68 USD InStock
TCM1221-025ML TCM1221-005ML
(+)-Menthyl chloroformate ≥97.0% (by titrimetric analysis)
(+)-Menthyl chloroformate
CAS Number: 7635-54-3
MDL Number: MFCD00134483
Molecular Formula: C11H19ClO2
Molecular Weight: 218.72
Purity/Analysis Method: >97.0% (T)
Form: Clear Liquid
Flash Point (°C): 70
Specific Gravity (20/20): 1.03
Specific rotation [a]20/D: 78 deg (neat)

Product Categories: Synthetic Organic Chemistry, Optical Resolving Agents, Optical Resolving Agents, Bioscience, Monocyclic Monoterpenes
Other Product Names / Synonyms: (+)-Chloroformyloxy-p-menthane


Formula: C₁₁H₁₉ClO₂
Density: 1.03 (20 °C)
Flash Pt: 70 °C
MDL Number: MFCD00134483
CAS Number: 7635-54-3
UN: 3277
ADR: 6.1,II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=26ac42bcc168643601725c5969a787713e41a1fa&vid=26ac42bcc168643601725c5969a787713e41a1fa&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >97.0% (T)

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty