Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-(-)-4-(Methoxymethyl)-1,3-dioxolan-2-one ≥98.0%

Supplier: Decon Laboratories Ltd

M1456-5G M1456-25G
TCM1456-5GEA 128 USD InStock
TCM1456-5G TCM1456-25G
(S)-(-)-4-(Methoxymethyl)-1,3-dioxolan-2-one ≥98.0%
(S)-(-)-4-(Methoxymethyl)-1,3-dioxolan-2-one
CAS Number: 135682-18-7
MDL Number: MFCD01321302
Molecular Formula: C5H8O4
Molecular Weight: 132.12
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 106
Flash Point (°C): 110
Specific Gravity (20/20): 1.24
Specific rotation [a]20/D: -46 deg (neat)

Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (S)-(-)-4-(Methoxymethyl)-2-oxo-1,3-dioxolane, (S)-(-)-3-Methoxypropylene Carbonate


Formula: C₅H₈O₄
Boiling Pt: 106 °C
Density: 1.24 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD01321302
CAS Number: 135682-18-7
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=56e78baddfbc6c72fb613f3083440f169ebe056e&vid=56e78baddfbc6c72fb613f3083440f169ebe056e&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty