Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Aminothiomorpholine-1,1-dioxide ≥95.0%

Supplier: Decon Laboratories Ltd

A1798-1G A1798-5G
TCA1798-1GEA 107 USD InStock
TCA1798-1G TCA1798-5G
4-Aminothiomorpholine-1,1-dioxide ≥95.0%
4-Aminothiomorpholine-1,1-dioxide
CAS Number: 26494-76-8
Molecular Formula: C4H10N2O2S
Molecular Weight: 150.20
Purity/Analysis Method: >95.0% (GC,T)
Form: Crystal
Melting point (°C): 107

Other Product Names / Synonyms: 1,1-Dioxo-4-thiomorpholinamine, 4-Thiomorpholinamine 1,1-Dioxide


Formula: C₄H₁₀N₂O₂S
MW: 150.20 g/mol
Melting Pt: 107 °C
CAS Number: 26494-76-8
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fce843c9597264df62c721bfe67eb3166d7fc43b&vid=fce843c9597264df62c721bfe67eb3166d7fc43b&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >95.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty