Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Megestrol acetate ≥96.0%

Supplier: Decon Laboratories Ltd

M1949-1G M1949-5G
TCM1949-1GEA 94 USD InStock
TCM1949-1G TCM1949-5G
Megestrol acetate ≥96.0%
Megestrol acetate
CAS Number: 595-33-5
MDL Number: MFCD00056470
Molecular Formula: C24H32O4
Molecular Weight: 384.52
Purity/Analysis Method: >96.0% (HPLC)
Form: Crystal
Melting point (°C): 218
Specific rotation [a]20/D: 11 deg (C=2, CHCl3)

Product Categories: Bioscience, Antitumor Ingredients, Pregnanes (Progestogen), Steroids
Other Product Names / Synonyms: Megestrol 17-Acetate, 17alpha-Acethoxy-6-methyl-4,6-pregnadiene-3,20-dione, 17alpha-Hydroxy-6-methyl-4,6-pregnadiene-3,20-dione 17-Acetate


Formula: C₂₄H₃₂O₄
MW: 384.52 g/mol
Melting Pt: 218 °C
MDL Number: MFCD00056470
CAS Number: 595-33-5
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fade2934a22da97ab561395d770812f0e9f5b94c&vid=fade2934a22da97ab561395d770812f0e9f5b94c&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >96.0% (HPLC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty