M1961-5G
M1961-1G
TCM1961-5GEA
195
USD
InStock
TCM1961-5G
TCM1961-1G
1-Methyl-N-carbobenzoxy-L-glutamate ≥98.0%
1-Methyl-N-carbobenzoxy-L-glutamate
CAS Number: 5672-83-3
MDL Number: MFCD00083278
Molecular Formula: C14H17NO6
Molecular Weight: 295.29
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 71
Specific rotation [a]20/D: -25 deg (C=1, EtOH)
Product Categories: Bioscience, Amino Acids (N-Protected), Amino Acids (C-Protected)
Other Product Names / Synonyms: N-Carbobenzoxy-L-glutamic Acid 1-Methyl Ester, 1-Methyl N-Cbz-L-glutamate, N-Cbz-L-glutamic Acid 1-Methyl Ester, Z-Glu-OMe