Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Methyl-1,2-epoxycyclohexane ≥95.0%

Supplier: Decon Laboratories Ltd

M1983-1G M1983-5G
TCM1983-1GEA 126 USD InStock
TCM1983-1G TCM1983-5G
1-Methyl-1,2-epoxycyclohexane ≥95.0%
1-Methyl-1,2-epoxycyclohexane
CAS Number: 1713-33-3
Molecular Formula: C7H12O
Molecular Weight: 112.17
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 138
Specific Gravity (20/20): 0.93

Other Product Names / Synonyms: 1,2-Epoxy-1-methylcyclohexane, 1-Methyl-7-oxabicyclo[4.1.0]heptane


Formula: C₇H₁₂O
Boiling Pt: 138 °C
Density: 0.93 (20 °C)
MDL Number: MFCD11846121
CAS Number: 1713-33-3
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=39ff5f69ec31ddfe3217cc103caf07fded0dbad2&vid=39ff5f69ec31ddfe3217cc103caf07fded0dbad2&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >95.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty