Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methyl-4'-(methylthio)-2-morpholinopropiophenone ≥98.0% (by GC, titration analysis)

Supplier: Decon Laboratories Ltd

M2028-500G M2028-25G
TCM2028-500GEA 374 USD InStock
TCM2028-500G TCM2028-25G
2-Methyl-4'-(methylthio)-2-morpholinopropiophenone ≥98.0% (by GC, titration analysis)
2-Methyl-4'-(methylthio)-2-morpholinopropiophenone
CAS Number: 71868-10-5
MDL Number: MFCD00083014
Molecular Formula: C15H21NO2S
Molecular Weight: 279.40
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 210
Melting point (°C): 75

Product Categories: Materials Chemistry, Polymerization Initiators
Other Product Names / Synonyms: 2-Methyl-1-[4-(methylthio)phenyl]-2-morpholino-1-propanone


Formula: C₁₅H₂₁NO₂S
Boiling Pt: 210 °C
Melting Pt: 75 °C
MDL Number: MFCD00083014
CAS Number: 71868-10-5
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0ff6430dfcece8e39f5626ab13058fad80c49322&vid=0ff6430dfcece8e39f5626ab13058fad80c49322&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty