Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(Mercaptomethyl)cyclopropaneacetic acid ≥97.0% (by GC, titration analysis)

Supplier: Decon Laboratories Ltd

M2074-1G M2074-5G
TCM2074-1GEA 83 USD InStock
TCM2074-1G TCM2074-5G
1-(Mercaptomethyl)cyclopropaneacetic acid ≥97.0% (by GC, titration analysis)
1-(Mercaptomethyl)cyclopropaneacetic acid
CAS Number: 162515-68-6
MDL Number: MFCD03411600
Molecular Formula: C6H10O2S
Molecular Weight: 146.20
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 45
Flash Point (°C): 110


Formula: C₆H₁₀O₂S
Melting Pt: 45 °C
Flash Pt: 110 °C
MDL Number: MFCD03411600
CAS Number: 162515-68-6
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=029daf40ba8284f7a9be120743cf3497cdec3379&vid=029daf40ba8284f7a9be120743cf3497cdec3379&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >97.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty