Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methoxyisonicotinic acid ≥98.0%

Supplier: Decon Laboratories Ltd

M2079-1G M2079-5G
TCM2079-1GEA 32.4 USD InStock
TCM2079-1G TCM2079-5G
2-Methoxyisonicotinic acid ≥98.0%
2-Methoxyisonicotinic acid
CAS Number: 105596-63-2
MDL Number: MFCD04115718
Molecular Formula: C7H7NO3
Molecular Weight: 153.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 203

Other Product Names / Synonyms: 2-Methoxypyridine-4-carboxylic Acid


Formula: C₇H₇NO₃
MW: 153.1400 g/mol
Melting Pt: 203 °C
MDL Number: MFCD04115718
CAS Number: 105596-63-2
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1bd381cd375da4e5bb1949f691bd615a38c90f45&vid=1bd381cd375da4e5bb1949f691bd615a38c90f45&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty