Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Nitrobenzenesulfonic acid hydrate ≥98.0% (by HPLC, titration analysis)

Supplier: Decon Laboratories Ltd

N0140-5G N0140-25G
TCN0140-005GEA 73 USD InStock
TCN0140-005G TCN0140-025G
4-Nitrobenzenesulfonic acid hydrate ≥98.0% (by HPLC, titration analysis)
4-Nitrobenzenesulfonic acid hydrate
CAS Number: 138-42-1
MDL Number: MFCD00041886
Molecular Formula: C6H5NO5S
Molecular Weight: 203.17
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 95


Formula: C₆H₅NO₅S
Melting Pt: 95 °C
MDL Number: MFCD00041886
CAS Number: 138-42-1
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6272f333369009be17f9d9b6ac031f9b69d5475&vid=f6272f333369009be17f9d9b6ac031f9b69d5475&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (HPLC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty