Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Nitrophenylhydrazine hydrochloride ≥98.0% for synthesis

Supplier: Decon Laboratories Ltd

N0233-25G N0233-5G
TCN0233-025GEA 159 USD InStock
TCN0233-025G TCN0233-5G
4-Nitrophenylhydrazine hydrochloride ≥98.0% for synthesis
4-Nitrophenylhydrazine hydrochloride
CAS Number: 636-99-7 MDL Number: MFCD00040592 Molecular Formula: C6H7N3O2 Molecular Weight: 189.60 Purity/Analysis Method: 98.0% (HPLC,T) Form: Crystal


Formula: C₆H₇N₃O₂·HCl
MW: 189.6 g/mol
MDL Number: MFCD00040592
CAS Number: 636-99-7
EINECS: 211-273-2
UN: 2811
ADR: 6.1 III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0c774df6218013d6a12c514f73a38bd35122263e&vid=0c774df6218013d6a12c514f73a38bd35122263e&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (HPLC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty