Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Nonadecanoic acid methyl ester ≥98.0%

Supplier: Decon Laboratories Ltd

N0460-5G N0460-25G
TCN0460-005GEA 175.44 USD InStock
TCN0460-005G TCN0460-025G
Nonadecanoic acid methyl ester ≥98.0%
Nonadecanoic acid methyl ester
CAS Number: 1731-94-8
MDL Number: MFCD00009011
Molecular Formula: C20H40O2
Molecular Weight: 312.54
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 40
Flash Point (°C): 113
Storage Temperature: 0-10°C

Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: Nonadecanoic Acid Methyl Ester


Formula: C₂₀H₄₀O₂
Melting Pt: 40 °C
Flash Pt: 113 °C
MDL Number: MFCD00009011
CAS Number: 1731-94-8
EINECS: 217-056-9
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e123c8f9832315058830ffbfeb51683ed07ec2cb&vid=e123c8f9832315058830ffbfeb51683ed07ec2cb&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty