Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Nonadecanedioic acid ≥98.0% (by titrimetric analysis)

Supplier: Decon Laboratories Ltd

N0663-5G N0663-1G
TCN0663-5GEA 902.8 USD InStock
TCN0663-5G TCN0663-1G
Nonadecanedioic acid ≥98.0% (by titrimetric analysis)
Nonadecanedioic acid
CAS Number: 6250-70-0
MDL Number: MFCD00039536
Molecular Formula: C19H36O4
Molecular Weight: 328.49
Purity/Analysis Method: >98.0% (T)
Form: Crystal

Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,17-Heptadecanedicarboxylic Acid


Formula: C₁₉H₃₆O₄
MDL Number: MFCD00039536
CAS Number: 6250-70-0
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=adfe847efdab32d94376bccaadcaab600eb9f44f&vid=adfe847efdab32d94376bccaadcaab600eb9f44f&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (T)

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty