Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Octadecylsuccinic anhydride ≥95.0% (by titrimetric analysis)

Supplier: Decon Laboratories Ltd

O0018-250G O0018-25G
TCO0018-250GEA 430 USD InStock
TCO0018-250G TCO0018-025G
Octadecylsuccinic anhydride ≥95.0% (by titrimetric analysis)
Octadecylsuccinic anhydride
CAS Number: 47458-32-2 MDL Number: MFCD00014551 Molecular Formula: C22H40O3 Molecular Weight: 352.56 Purity/Analysis Method: 95.0% (T) Form: Crystal Melting point (°C): 79


Formula: C₂₂H₄₀O₃
Melting Pt: 79 °C
MDL Number: MFCD00014551
CAS Number: 47458-32-2
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ef6bf6f92edb4babae930b5597da1282a86cb145&vid=ef6bf6f92edb4babae930b5597da1282a86cb145&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method 95.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty