Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Oleamide 65.0%

Supplier: Decon Laboratories Ltd

O0107-500G O0107-25G
TCO0107-500GEA 116 USD InStock
TCO0107-500G TCO0107-025G
Oleamide 65.0%
Oleamide
CAS Number: 301-02-0
MDL Number: MFCD00053638
Molecular Formula: C18H35NO
Molecular Weight: 281.48
Purity/Analysis Method: >65.0% (GC)
Form: Crystal
Melting point (°C): 70
Flash Point (°C): 216

Product Categories: Materials Chemistry, Pressure & Heat Sensitive Recording Materials
Other Product Names / Synonyms: cis-9-Octadecenamide, Oleyramide


Formula: C₁₈H₃₅NO
MDL Number: MFCD00053638
CAS Number: 301-02-0
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=115e818c27b63c838d669202ea735c00ed4f2c80&vid=115e818c27b63c838d669202ea735c00ed4f2c80&prid=web&clid=SAPDAM
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty