Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium 1-octadecanesulfonate

Supplier: Decon Laboratories Ltd

O0124-25G O0124-5G
TCO0124-25GEA 484 USD InStock
TCO0124-25G TCO0124-005G
Sodium 1-octadecanesulfonate
Sodium 1-octadecanesulphonate
CAS Number: 13893-34-0
MDL Number: MFCD00051526
Molecular Formula: C18H38O3S
Molecular Weight: 356.54
Form: Crystal
Color: White

Other Product Names / Synonyms: 1-Octadecanesulfonic Acid Sodium Salt, Octadecylsulfonic Acid Sodium Salt


Formula: C₁₈H₃₇NaO₃S
MDL Number: MFCD00051526
CAS Number: 13893-34-0
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=819dfc50642673f13096a88d0001a447f802df95&vid=819dfc50642673f13096a88d0001a447f802df95&prid=web&clid=SAPDAM
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty