Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2'-Oxybis(ethylamine) ≥98.0% (by GC, titration analysis)

Supplier: Decon Laboratories Ltd

O0278-1G O0278-25G O0278-5G
TCO0278-001GEA 262 USD InStock
TCO0278-001G TCO0278-25G TCO0278-5G
2,2'-Oxybis(ethylamine) ≥98.0% (by GC, titration analysis)
2,2'-Oxybis(ethylamine)
CAS Number: 2752-17-2
MDL Number: MFCD00466591
Molecular Formula: C4H12N2O
Molecular Weight: 104.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 64
Specific Gravity (20/20): 0.98

Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Bis(2-aminoethyl) Ether, 2,2'-Diaminodiethyl Ether, 2,2'-Oxydiethylamine


Formula: C₄H₁₂N₂O
Boiling Pt: 64 °C
Density: 0.98 (20 °C)
MDL Number: MFCD00466591
CAS Number: 2752-17-2
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ac43b8e130345eec00e9f3f8ca62469f73a2e19&vid=8ac43b8e130345eec00e9f3f8ca62469f73a2e19&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty