Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Phenylazobenzoyl chloride ≥98.0%

Supplier: Decon Laboratories Ltd

P0144-5G P0144-1G
TCP0144-005GEA 329 USD InStock
TCP0144-005G TCP0144-001G
4-Phenylazobenzoyl chloride ≥98.0%
4-Phenylazobenzoyl chloride
CAS Number: 104-24-5
MDL Number: MFCD00041722
Molecular Formula: C13H9ClN2O
Molecular Weight: 244.68
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 95

Other Product Names / Synonyms: Azobenzene-4-carbonyl Chloride, Azoyl Chloride


Formula: C₁₃H₉ClN₂O
Melting Pt: 95 °C
MDL Number: MFCD00041722
CAS Number: 104-24-5
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d2749785a7cc4eba1bd405be0569184e360ae259&vid=d2749785a7cc4eba1bd405be0569184e360ae259&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty