Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1S)-(-)-β-Pinene ≥94.0%

Supplier: Decon Laboratories Ltd

P0441-25ML P0441-500ML
TCP0441-025MLEA 23.4 USD InStock
TCP0441-025ML TCP0441-500ML
(1S)-(-)-β-Pinene ≥94.0%
(1S)-(-)-β-Pinene
CAS Number: 18172-67-3
MDL Number: MFCD00063635
Molecular Formula: C10H16
Molecular Weight: 136.24
Purity/Analysis Method: >94.0% (GC)
Form: Clear Liquid
Boiling point (°C): 165
Flash Point (°C): 43
Specific Gravity (20/20): 0.87
Specific rotation [a]20/D: -21 deg (neat)


Formula: C₁₀H₁₆
Boiling Pt: 165 °C
Melting Pt: –61 °C
Density: 0.87 (20 °C)
Flash Pt: 43 °C
MDL Number: MFCD00001345
CAS Number: 18172-67-3
EINECS: 242-060-2
UN: 2319
ADR: 3,III
Merck Index: 12,07600
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=39690ebc5db711501a0dc659b54f4b6d4f97871c&vid=39690ebc5db711501a0dc659b54f4b6d4f97871c&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >94.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty