Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pregnenolone acetate ≥99.0% (by HPLC)

Supplier: Decon Laboratories Ltd

P0477-10G P0477-1G
TCP0477-010GEA 98 USD InStock
TCP0477-010G TCP0477-001G
Pregnenolone acetate ≥99.0% (by HPLC)
Pregnenolone acetate
CAS Number: 1778-02-5 MDL Number: MFCD00003627 Molecular Formula: C23H34O3 Molecular Weight: 358.52 Purity/Analysis Method: 99.0% (HPLC) Form: Crystal Melting point (°C): 151 Specific rotation [a]20/D: 20 deg (C=1, EtOH)


Formula: C₂₃H₃₄O₃
Melting Pt: 151 °C
MDL Number: MFCD00003627
CAS Number: 1778-02-5
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b67126e3a4a5d18f870398b4b9f3063f3d0f5adf&vid=b67126e3a4a5d18f870398b4b9f3063f3d0f5adf&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method 99.0% (HPLC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty