Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N,N'-1,3-Phenylenedimaleimide ≥97.0%

Supplier: Decon Laboratories Ltd

P0976-25G P0976-500G
TCP0976-025GEA 52.8 USD InStock
TCP0976-025G TCP0976-500G
N,N'-1,3-Phenylenedimaleimide ≥97.0%
N,N'-1,3-Phenylenedimaleimide
CAS Number: 3006-93-7
MDL Number: MFCD00022569
Molecular Formula: C14H8N2O4
Molecular Weight: 268.23
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Color: Pale Yellow
Melting point (°C): 202
Flash Point (°C): 208

Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: N,N'-1,3-Bismaleimidobenzene, N,N'-1,3-Phenylenebismaleimide


Formula: C₁₄H₈N₂O₄
Melting Pt: 202 °C
Flash Pt: 208 °C
MDL Number: MFCD00022569
CAS Number: 3006-93-7
Order Now
Specification Test Results
Purity/Analysis Method >97.0% (GC)

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty