Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Pyrenemethanol ≥98.0%

Supplier: Decon Laboratories Ltd

P1221-1G P1221-5G
TCP1221-001GEA 45 USD InStock
TCP1221-001G TCP1221-5G
1-Pyrenemethanol ≥98.0%
1-Pyrenemethanol
CAS Number: 24463-15-8
MDL Number: MFCD00029252
Molecular Formula: C17H12O
Molecular Weight: 232.28
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 125

Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors
Other Product Names / Synonyms: 1-(Hydroxymethyl)pyrene


Formula: C₁₇H₁₂O
Melting Pt: 125 °C
MDL Number: MFCD00029252
CAS Number: 24463-15-8
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5d747af7724fe47de8678feb45d84d98f6bf754e&vid=5d747af7724fe47de8678feb45d84d98f6bf754e&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty