P1234-25G
P1234-500G
TCP1234-025GEA
46
USD
InStock
TCP1234-025G
TCP1234-500G
Palatinose hydrate ≥98.0% (by HPLC)
Palatinose hydrate
CAS Number: 13718-94-0
MDL Number: MFCD00076094
Molecular Formula: C12H22O11
Molecular Weight: 342.30
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 123
Specific rotation [a]20/D: 99 deg (C=1, H2O)
Product Categories: Bioscience, Obesity/Diabete Research Reagents, Basic Sugars (Mono & Oligosaccharides), Disaccharides
Other Product Names / Synonyms: 6-O-alpha-D-Glucopyranosyl-D-fructose, Isomaltulose