Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Azacyclonol ≥99.0% (by GC, titration analysis)

Supplier: Decon Laboratories Ltd

P1369-25G P1369-5G
TCP1369-025GEA 124 USD InStock
TCP1369-025G TCP1369-5G
Azacyclonol ≥99.0% (by GC, titration analysis)
Azacyclonol
CAS Number: 115-46-8
MDL Number: MFCD00066980
Molecular Formula: C18H21NO
Molecular Weight: 267.37
Purity/Analysis Method: >99.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 163

Other Product Names / Synonyms: alpha,alpha-Diphenyl-4-piperidinemethanol


Formula: C₁₈H₂₁NO
Melting Pt: 163 °C
MDL Number: MFCD00066980
CAS Number: 115-46-8
EINECS: 204-092-5
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=18f9d3078675a1c04703afa5cf2085a864263ea1&vid=18f9d3078675a1c04703afa5cf2085a864263ea1&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >99.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty