Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Propyl methanesulfonate ≥98.0%

Supplier: Decon Laboratories Ltd

P1844-25G P1844-5G
TCP1844-25GEA 402 USD InStock
TCP1844-25G TCP1844-5G
Propyl methanesulfonate ≥98.0%
Propyl methanesulfonate
CAS Number: 1912-31-8
MDL Number: MFCD00007561
Molecular Formula: C4H10O3S
Molecular Weight: 138.18
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 110
Specific Gravity (20/20): 1.15

Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives
Other Product Names / Synonyms: Methanesulfonic Acid Propyl Ester


Formula: C₄H₁₀O₃S
Boiling Pt: 110 °C
Density: 1.15 (20 °C)
MDL Number: MFCD00007561
CAS Number: 1912-31-8
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c87448cac4f1bea422d81b263cbc80e70d594308&vid=c87448cac4f1bea422d81b263cbc80e70d594308&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty