S0087-500G
S0087-25G
S0087-100G
TCS0087-500GEA
415
USD
InStock
TCS0087-500G
TCS0087-025G
TCS0087-100G
Trimethylstearylammonium chloride ≥98.0%
Trimethylstearylammonium chloride
CAS Number: 112-03-8
MDL Number: MFCD00050188
Molecular Formula: C21H46ClN
Molecular Weight: 348.06
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Product Categories: Materials Chemistry, Cationic Surfactants
Other Product Names / Synonyms: Octadecyltrimethylammonium Chloride, Trimethyloctadecylammonium Chloride, STAC