Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

β-Sitosterol acetate 40,0% (contains Campesterol Acetate)

Supplier: Decon Laboratories Ltd

S0221-1G S0221-5G
TCS0221-001GEA 108 USD InStock
TCS0221-001G TCS0221-5G
β-Sitosterol acetate 40,0% (contains Campesterol Acetate)
beta-Sitosterol acetate
CAS Number: 915-05-9
MDL Number: MFCD00083497
Molecular Formula: C31H52O2
Molecular Weight: 456.76
Purity/Analysis Method: >40.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 133


Formula: C₃₁H₅₂O₂
Melting Pt: 133 °C
MDL Number: MFCD00083497
CAS Number: 915-05-9
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bedf9954fe683eece4d6ae1d2697a1dcd0b2a9aa&vid=bedf9954fe683eece4d6ae1d2697a1dcd0b2a9aa&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >40.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty