Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Scopolamine methyl nitrate ≥98.0% (by HPLC, titration analysis)

Supplier: Decon Laboratories Ltd

S0230-1G S0230-5G
TCS0230-001GEA 160.72 USD InStock
TCS0230-001G TCS0230-005G
Scopolamine methyl nitrate ≥98.0% (by HPLC, titration analysis)
Scopolamine methyl nitrate
CAS Number: 6106-46-3
MDL Number: MFCD00078239
Molecular Formula: C18H24N2O7
Molecular Weight: 380.40
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 199
Specific rotation [a]20/D: -25 deg (C=1, H2O)

Product Categories: Bioscience, Neurotransmitter Agent Ingredients, Acetylcholine Receptor (AChR) Agent Ingredients, Tropane Alkaloids
Other Product Names / Synonyms: Methscopolamine Nitrate


Formula: C₁₈H₂₄N₂O₇
MDL Number: MFCD00078239
CAS Number: 6106-46-3
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ff2694ecc7814fe3b8728ca12e29eec3abcecc6&vid=8ff2694ecc7814fe3b8728ca12e29eec3abcecc6&prid=web&clid=SAPDAM
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty