Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Salicylideneaminophenol ≥98.0% (by titrimetric analysis)

Supplier: Decon Laboratories Ltd

S0328-5G S0328-25G
TCS0328-005GEA 51.3 USD InStock
TCS0328-005G TCS0328-025G
2-Salicylideneaminophenol ≥98.0% (by titrimetric analysis)
2-Salicylideneaminophenol
CAS Number: 1761-56-4
MDL Number: MFCD00020011
Molecular Formula: C13H11NO2
Molecular Weight: 213.24
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: Reddish Yellow
Melting point (°C): 187


Formula: C₁₃H₁₁NO₂
Melting Pt: 187 °C
MDL Number: MFCD00020011
CAS Number: 1761-56-4
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f9fc2e6f17586c9fcff3063f5449bb2a9a225099&vid=f9fc2e6f17586c9fcff3063f5449bb2a9a225099&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty