Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Artemether ≥98.0% (by HPLC)

Supplier: Decon Laboratories Ltd

A2190-5G A2190-25G
TCA2190-5GEA 81 USD InStock
TCA2190-5G TCA2190-25G
Artemether ≥98.0% (by HPLC)
Artemether
CAS Number: 71963-77-4
MDL Number: MFCD00866205
Molecular Formula: C16H26O5
Molecular Weight: 298.38
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 89
Specific rotation [a]20/D: 170 deg (C=1, EtOH)


Formula: C₁₆H₂₆O₅
Melting Pt: 89 °C
MDL Number: MFCD00866205
CAS Number: 71963-77-4
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9291ed9df55dccf1800eaf7d933fdab98a7f21c2&vid=9291ed9df55dccf1800eaf7d933fdab98a7f21c2&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (HPLC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty