Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-4-(chloroformyl)benzoate ≥95.0%

Supplier: Decon Laboratories Ltd

T0283-500G T0283-25G
TCT0283-500GEA 1002 USD InStock
TCT0283-500G TCT0283-025G
Methyl-4-(chloroformyl)benzoate ≥95.0%
Methyl-4-(chloroformyl)benzoate
CAS Number: 7377-26-6
MDL Number: MFCD00059486
Molecular Formula: C9H7ClO3
Molecular Weight: 198.60
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 140
Melting point (°C): 54

Other Product Names / Synonyms: 4-(Chloroformyl)benzoic Acid Methyl Ester, Terephthalic Acid Monomethyl Ester Chloride, Methyl 4-(Chlorocarbonyl)benzoate


Formula: C₉H₇ClO₃
Boiling Pt: 140 °C
Melting Pt: 54 °C
MDL Number: MFCD00059486
CAS Number: 7377-26-6
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7a9610481b3a198b98e95522d1133b6f7d620943&vid=7a9610481b3a198b98e95522d1133b6f7d620943&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >95.0% (GC)

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty