Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(2-Aminoethyl)cyclohexanol ≥98.0%

Supplier: Decon Laboratories Ltd

A2266-5G A2266-1G
TCA2266-5GEA 718 USD InStock
TCA2266-5G TCA2266-1G
4-(2-Aminoethyl)cyclohexanol ≥98.0%
4-(2-Aminoethyl)cyclohexanol
CAS Number: 148356-06-3
Molecular Formula: C8H17NO
Molecular Weight: 143.23
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 256


Formula: C₈H₁₇NO
Boiling Pt: 256 °C
MDL Number: MFCD11520548
CAS Number: 148356-06-3
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9d5371d8ebc63d1fb69496d354f407e4740a28b5&vid=9d5371d8ebc63d1fb69496d354f407e4740a28b5&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty