Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tetrakis(hydroxymethyl)phosphonium sulfate ∼70 - 80% in water

Supplier: Decon Laboratories Ltd

T1089-25G T1089-500G
TCT1089-025GEA 30 USD InStock
TCT1089-025G TCT1089-500G
Tetrakis(hydroxymethyl)phosphonium sulfate ∼70 - 80% in water
Tetrakis(hydroxymethyl)phosphonium sulfate
CAS Number: 55566-30-8
MDL Number: MFCD00054243
Molecular Formula: C4H12O4P
Molecular Weight: 406.28
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 96


Formula: C₄H₁₂O₄P
MW: 406.28 g/mol
Flash Pt: 96 °C
MDL Number: MFCD00054243
CAS Number: 55566-30-8
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2be52a929a6456a2679b3caeff395361e1af3494&vid=2be52a929a6456a2679b3caeff395361e1af3494&prid=web&clid=SAPDAM
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty