Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Copper(I) trifluoromethanesulfonate benzene complex ≥90.0% (by titrimetric analysis)

Supplier: Decon Laboratories Ltd
Warning

T1442-1G T1442-5G
TCT1442-001GEA 72.7 USD InStock
TCT1442-001G TCT1442-5G
Copper(I) trifluoromethanesulfonate benzene complex ≥90.0% (by titrimetric analysis)
Copper(I) trifluoromethanesulfonate benzene complex
CAS Number: 42152-46-5 MDL Number: MFCD00043300 Molecular Formula: CCuF3O3S Molecular Weight: 251.67 Purity/Analysis Method: 90.0% (T) Form: Crystal Color: White Melting point (°C): 160 Flash Point (°C): 7 Storage Temperature: 0-10°C

Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Homogeneous Catalysts, Cross-coupling Reaction using Transition Metal Catalysts, Metal Catalysts & Ligands (Click Chemistry)
Other Product Names / Synonyms: Copper(I) Triflate Benzene Complex, Trifluoromethanesulfonic Acid Copper(I) Salt Benzene Complex


Formula: CCuF₃O₃S
MW: 503.33 g/mol
Melting Pt: 160 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00043300
CAS Number: 42152-46-5
EINECS: 255-686-6
UN: 3175
ADR: 4.1 II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47ca99f918e6f71dcc36c2989dff78ab0815c53b&vid=47ca99f918e6f71dcc36c2989dff78ab0815c53b&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >90.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty